Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Difluorobenzaldehyde 97%

2,6-Difluorobenzaldehyde 97%

Bottle, 100 g
Catalog # APOSPC2615D-100G
Irritant
Supplier:   Apollo Scientific
2,6-Difluorobenzaldehyde 97%
Catalog # APOSPC2615D-100G
Supplier:  Apollo Scientific
CAS Number:  437-81-0
2,6-Difluorobenzaldehyde 97%

Bottle, 100 g

CAS Number:  437-81-0
Price
123,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:437-81-0
  • Formula:C₇H₄F₂O
  • MDL Number:MFCD00010293
  • Molecular Weight:142.1 g/mol
  • Boiling Pt:82…84 °C/15mmHg
  • Melting Pt:15…17 °C
  • Density:1.317 g/cm³ (25 °C)
  • Flash Pt:73 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications