Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Maltose monohydrate

D-(+)-Maltose monohydrate

Bottle, 1 kg
Catalog # APOSBIK9000-1KG
Irritant
Supplier:   Apollo Scientific
D-(+)-Maltose monohydrate
Catalog # APOSBIK9000-1KG
Supplier:  Apollo Scientific
CAS Number:  6363-53-7
D-(+)-Maltose monohydrate

Bottle, 1 kg

CAS Number:  6363-53-7
Price
79,60 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
  • CAS Number:6363-53-7
  • Formula:C₁₂H₂₂O₁₁·1H₂O
  • MDL Number:MFCD00149343
  • Molecular Weight:360.32 g/mol
  • Melting Pt:120 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-716-5
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications