Order Entry
Export
Orders LinkContactUsLinkComponent
Patulin standard solution

Patulin standard solution

Patulin, 100 µg/ml in acetonitrile, Bottle, 1 mL
Catalog # APOSBIMS108-1ML
Supplier:   Apollo Scientific
Patulin standard solution
Catalog # APOSBIMS108-1ML
Supplier:  Apollo Scientific
CAS Number:  
Patulin standard solution

Patulin, 100 µg/ml in acetonitrile, Bottle, 1 mL

CAS Number:  
Price
218,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Analytical reference standard solution in acetonitrile.

Caution: Expires in 12 months.

Documents

Specifications

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pack type:
    Bottle
  • Pk:
    1 mL
  • Description:
    Patulin
  • Cat. No.:
    APOSBIMS108-1ML
  • Formula:
    C₇H₆O₄
  • Description:
    Patulin
  • Boiling point:
    81…82 °C
  • Storage temperature:
    Freezer
  • Flash point:
    2 °C
  • CAS No.:
    149-29-1

Specifications