Fumonisin Standard Solutions
Fumonisin B1 and Fumonisin B2 mixed, 50 µg/ml of each in ACN/H2O, 5 mL
Catalog # APOSBIMS102-5ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Analytical reference standard solutions derived from Fusarium moniliforme. Also known as Macrofusine
- 50µg/ml of each component in ACN/H2O 1:1 V/V mixture.
Carcinogenic mycotoxin.
Caution: Expires in 12 months.
Specifications
Specifications
- Concentration:50 µg/ml of each in ACN/H2O
- Pk:5 mL
- Description:Fumonisin B1 and Fumonisin B2 mixed



