Order Entry
Export
Orders LinkContactUsLinkComponent
Fumonisin Standard Solutions

Fumonisin Standard Solutions

Fumonisin B1 and Fumonisin B2 mixed, 50 µg/ml of each in ACN/H2O, 1 mL
Catalog # APOSBIMS102-1ML
Supplier:   Apollo Scientific
Fumonisin Standard Solutions
Catalog # APOSBIMS102-1ML
Supplier:  Apollo Scientific
CAS Number:  
Fumonisin Standard Solutions

Fumonisin B1 and Fumonisin B2 mixed, 50 µg/ml of each in ACN/H2O, 1 mL

CAS Number:  
Price
290,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Analytical reference standard solutions derived from Fusarium moniliforme. Also known as Macrofusine

  • 50µg/ml of each component in ACN/H2O 1:1 V/V mixture.

Carcinogenic mycotoxin.

Caution: Expires in 12 months.

Specifications

Specifications

  • Concentration:
    50 µg/ml of each in ACN/H2O
  • Pk:
    1 mL
  • Description:
    Fumonisin B1 and Fumonisin B2 mixed

Specifications