Order Entry
Export
Orders LinkContactUsLinkComponent
Fumonisin Standard Solutions

Fumonisin Standard Solutions

Fumonisin B2, 50 µg/ml in ACN/H2O, 5 mL
Catalog # APOSBIMS101-5ML
Supplier:   Apollo Scientific
Fumonisin Standard Solutions
Catalog # APOSBIMS101-5ML
Supplier:  Apollo Scientific
CAS Number:  
Fumonisin Standard Solutions

Fumonisin B2, 50 µg/ml in ACN/H2O, 5 mL

CAS Number:  
Price
992,00 €
Log in for availability
1 * 5 mL
Product Details & Documents

Analytical reference standard solutions derived from Fusarium moniliforme. Also known as Macrofusine

  • 50µg/ml of each component in ACN/H2O 1:1 V/V mixture.

Carcinogenic mycotoxin.

Caution: Expires in 12 months.

Specifications

Specifications

  • Concentration:
    50 µg/ml in ACN/H2O
  • Pk:
    5 mL
  • Description:
    Fumonisin B2
  • Cat. No.:
    APOSBIMS101-5ML
  • Formula:
    C₃₄H₅₉NO₁₄
  • Description:
    Fumonisin B2
  • Storage temperature:
    Freezer
  • CAS No.:
    116355-84-1

Specifications