Fumonisin Standard Solutions
Fumonisin B2, 50 µg/ml in ACN/H2O, 1 mL
Catalog # APOSBIMS101-1ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Analytical reference standard solutions derived from Fusarium moniliforme. Also known as Macrofusine
- 50µg/ml of each component in ACN/H2O 1:1 V/V mixture.
Carcinogenic mycotoxin.
: Expires in 12 months.
Specifications
Specifications
- Concentration:50 µg/ml in ACN/H2O
- Pk:1 mL
- Description:Fumonisin B2
- Cat. No.:APOSBIMS101-1ML
- Formula:C₃₄H₅₉NO₁₄
- Description:Fumonisin B2
- Storage temperature:Freezer
- CAS No.:116355-84-1



