Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fumonisin Standard Solutions

Fumonisin B1, 50 µg/ml in ACN/H2O, 5 mL
  APOSBIMS100-5ML
Fumonisin Standard Solutions
  APOSBIMS100-5ML
 :  
Fumonisin Standard Solutions

Fumonisin B1, 50 µg/ml in ACN/H2O, 5 mL

 :  
 
992,00 €
   
1 * 5 mL
 

Analytical reference standard solutions derived from Fusarium moniliforme. Also known as Macrofusine

  • 50µg/ml of each component in ACN/H2O 1:1 V/V mixture.

Carcinogenic mycotoxin.

 : Expires in 12 months.

 

 

  • Concentration:
    50 µg/ml in ACN/H2O
  • Pk:
    5 mL
  • Description:
    Fumonisin B1
  • Cat. No.:
    APOSBIMS100-5ML
  • Formula:
    C₃₄H₅₉NO₁₅
  • Description:
    Fumonisin B1
  • Storage temperature:
    Freezer
  • CAS No.:
    116355-83-0