Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Difluoroaniline 98%

3,5-Difluoroaniline 98%

Bottle, 25 g
Catalog # APOSPC2613-25G
Toxic
Irritant
Health
Supplier:   Apollo Scientific
3,5-Difluoroaniline 98%
Catalog # APOSPC2613-25G
Supplier:  Apollo Scientific
CAS Number:  372-39-4
3,5-Difluoroaniline 98%

Bottle, 25 g

CAS Number:  372-39-4
Price
24,60 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:372-39-4
  • Formula:C₆H₅F₂N
  • MDL Number:MFCD00007763
  • Molecular Weight:129.11 g/mol
  • Boiling Pt:80 °C/20mmHg
  • Melting Pt:40…42 °C
  • Flash Pt:75 °C
  • Storage Temperature:Refrigerator
  • EINECS:206-752-8
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications