Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Difluorophenol 98%

2,4-Difluorophenol 98%

Bottle, 100 g
Catalog # APOSPC2606-100G
Flammable
Irritant
Supplier:   Apollo Scientific
2,4-Difluorophenol 98%
Catalog # APOSPC2606-100G
Supplier:  Apollo Scientific
CAS Number:  367-27-1
2,4-Difluorophenol 98%

Bottle, 100 g

CAS Number:  367-27-1
Price
75,30 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:367-27-1
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00009715
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:52…53 °C/19mmHg
  • Melting Pt:22 °C
  • Density:1.362 g/cm³ (25 °C)
  • Flash Pt:57 °C
  • Storage Temperature:Ambient
  • EINECS:206-688-0
  • UN:1993
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications