Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Difluoropropane 97%

Bottle, 5 g
Catalog # APOSPC2877B-5G
Irritant
Supplier:   Apollo Scientific
1,3-Difluoropropane 97%
Catalog # APOSPC2877B-5G
Supplier:  Apollo Scientific
CAS Number:  462-39-5
1,3-Difluoropropane 97%

Bottle, 5 g

CAS Number:  462-39-5
Price
267,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:462-39-5
  • Formula:C₃H₆F₂
  • MDL Number:MFCD00040969
  • Molecular Weight:80.08 g/mol
  • Boiling Pt:40…42 °C
  • Density:1.006 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications