0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Difluoropropane
1,3-Difluoropropane 97%
1,3-Difluoropropane 97%
1,3-Difluoropropane 97%
Bottle, 25 g
APOSPC2877B-25G
:
Apollo Scientific
1,3-Difluoropropane 97%
APOSPC2877B-25G
:
Apollo Scientific
:
462-39-5
Expand image
1,3-Difluoropropane 97%
Bottle, 25 g
Add to cart
:
462-39-5
436,00 €
Unit of Measure
1 * 25 g
-
+
Add to cart
:
462-39-5
:
C₃H₆F₂
:
MFCD00040969
:
80.08 g/mol
:
40…42 °C
:
1.006 g/cm³ (25 °C)
:
Refrigerator
Pack type:
Bottle
Pk:
25 g