Orders LinkContactUsLinkComponent
L(+)-Glutamine ≥99%
Bottle, 1 kg
  APOSBIG0708-1KG
L(+)-Glutamine ≥99%
  APOSBIG0708-1KG
 :  56-85-9
L(+)-Glutamine ≥99%
Bottle, 1 kg
 :  56-85-9
91,20 €
1 * 1 kg
Pyrogen free
 :  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  •  :56-85-9
  •  :C₅H₁₀N₂O₃
  •  :MFCD00008044
  •  :146.15 g/mol
  •  :185 °C (dec.)
  •  :Ambient
  •  :200-292-1
  • Pack type:
    Bottle
  • Pk:
    1 kg