Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Glutamine ≥99%

L(+)-Glutamine ≥99%

Bottle, 100 g
Catalog # APOSBIG0708-100G
Irritant
Supplier:   Apollo Scientific
L(+)-Glutamine ≥99%
Catalog # APOSBIG0708-100G
Supplier:  Apollo Scientific
CAS Number:  56-85-9
L(+)-Glutamine ≥99%

Bottle, 100 g

CAS Number:  56-85-9
Price
21,80 €
Log in for availability
1 * 100 g
Product Details & Documents

Pyrogen free

Synonyms:  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  • CAS Number:56-85-9
  • Formula:C₅H₁₀N₂O₃
  • MDL Number:MFCD00008044
  • Molecular Weight:146.15 g/mol
  • Melting Pt:185 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-292-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications