Order Entry
Export
Orders LinkContactUsLinkComponent
trans-1,2-Cyclohexanedicarboxylic acid

trans-1,2-Cyclohexanedicarboxylic acid

Bottle, 100 g
Catalog # APOSOR350372-100G
Irritant
Supplier:   Apollo Scientific
trans-1,2-Cyclohexanedicarboxylic acid
Catalog # APOSOR350372-100G
Supplier:  Apollo Scientific
CAS Number:  2305-32-0
trans-1,2-Cyclohexanedicarboxylic acid

Bottle, 100 g

CAS Number:  2305-32-0
Price
30,40 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  trans-Cyclohexane-1,2-dicarboxylic acid
  • CAS Number:2305-32-0
  • Formula:C₈H₁₂O₄
  • MDL Number:MFCD00001464
  • Molecular Weight:172.18 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications