Agarose
Bottle, 1 kg
Catalog # APOSBIA1176-1KG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- Gel Strength 1.5% >100g/cm2?2200g/cm2
- Gelling temp 36 ± 1.5C
- Melting temp 88 ± 1.5C
- Ash Content <0.45%
- Sulphate <0.1%
- Clarity <4 NTU
- DNA/RNAse Negative.
- Use Rates 0.75% - 2%.
- EEO 0.14 - 0.16
- CAS Number:9012-36-6
- MDL Number:MFCD00081294
- Storage Temperature:Ambient
- EINECS:232-731-8
Specifications
Specifications
- Pack type:Bottle
- Pk:1 kg



