Order Entry
Export
Orders LinkContactUsLinkComponent
Sesquicillin A

Sesquicillin A

Bottle, 5 mg
Catalog # APOSBIBR1155-5MG
Toxic
Supplier:   Apollo Scientific
Sesquicillin A
Catalog # APOSBIBR1155-5MG
Supplier:  Apollo Scientific
CAS Number:  51103-58-3
Sesquicillin A

Bottle, 5 mg

CAS Number:  51103-58-3
Price
1 060,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

From microbial source

  • CAS Number:51103-58-3
  • Formula:C₂₉H₄₂O₅
  • MDL Number:MFCD08274594
  • Molecular Weight:470.641 g/mol
  • Storage Temperature:Freezer
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 mg

Specifications