Cochlioquinone A
Bottle, 5 mg
Catalog # APOSBIBR1150-5MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
From microbial source. Purity: (c.70%)
Synonyms: Luteoleersin
- CAS Number:32450-25-2
- Formula:C₃₀H₄₄O₈
- MDL Number:MFCD08062299
- Molecular Weight:532.666 g/mol
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Bottle
- Pk:5 mg



