1,1-Dichloroethane 98%
Bottle, 5 g
Catalog # APOSOR3712-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Ethylidene dichloride
- CAS Number:75-34-3
- Formula:Cl₂HCCH₃
- MDL Number:MFCD00013673
- Molecular Weight:98.96 g/mol
- Boiling Pt:57 °C
- Melting Pt:–98 °C
- Density:1.18 g/cm³ (25 °C)
- Flash Pt:–10 °C
- Storage Temperature:Ambient
- EINECS:200-863-5
- UN:2362
- ADR:3,II
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g



