Order Entry
Export
Orders LinkContactUsLinkComponent
 

Taurine ≥99%

Bottle, 100 g
Catalog # APOSBIA0603-100G
Irritant
Supplier:   Apollo Scientific
Taurine ≥99%
Catalog # APOSBIA0603-100G
Supplier:  Apollo Scientific
CAS Number:  107-35-7
Taurine ≥99%

Bottle, 100 g

CAS Number:  107-35-7
Price
21,80 €
Log in for availability
1 * 100 g
Product Details & Documents
  • Water (by Karl Fischer) <1.0%
Synonyms:  2-Aminoethanesulphonic acid
  • CAS Number:107-35-7
  • Formula:C₂H₇NO₃S
  • MDL Number:MFCD00008197
  • Molecular Weight:125.15 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:203-483-8

 

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications