Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Difluorophenetole

2,3-Difluorophenetole

Bottle, 5 g
Catalog # APOSPC1345-5G
Irritant
Supplier:   Apollo Scientific
2,3-Difluorophenetole
Catalog # APOSPC1345-5G
Supplier:  Apollo Scientific
CAS Number:  121219-07-6
2,3-Difluorophenetole

Bottle, 5 g

CAS Number:  121219-07-6
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1-Ethoxy-2,3-difluorobenzene, 2,3-Difluoroethoxybenzene
  • CAS Number:121219-07-6
  • Formula:C₈H₈F₂O
  • MDL Number:MFCD07368737
  • Molecular Weight:158.15 g/mol
  • Boiling Pt:178 °C
  • Density:1.18 g/cm³ (25 °C)
  • Flash Pt:70.5 °C
  • Storage Temperature:Ambient
  • EINECS:441-000-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications