Order Entry
Export
Orders LinkContactUsLinkComponent
Bisacrylamide ≥98%

Bisacrylamide ≥98%

Bottle, 250 g
Catalog # APOSBIB6418-250G
Irritant
Supplier:   Apollo Scientific
Bisacrylamide ≥98%
Catalog # APOSBIB6418-250G
Supplier:  Apollo Scientific
CAS Number:  110-26-9
Bisacrylamide ≥98%

Bottle, 250 g

CAS Number:  110-26-9
Price
138,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-750-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications