Order Entry
Export
Orders LinkContactUsLinkComponent
Bis-trispropane 99+%

Bis-trispropane 99+%

Bottle, 500 g
Catalog # APOSBI6850-500G
Irritant
Supplier:   Apollo Scientific
Bis-trispropane 99+%
Catalog # APOSBI6850-500G
Supplier:  Apollo Scientific
CAS Number:  64431-96-5
Bis-trispropane 99+%

Bottle, 500 g

CAS Number:  64431-96-5
Price
254,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Biological buffer - useful range 6.3-9.5

  • Assay (by titration) 99+%
  • Water (by Karl Fischer) < 1.0%
Synonyms:  1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
  • CAS Number:64431-96-5
  • Formula:C₁₁H₂₆N₂O₆
  • MDL Number:MFCD00004689
  • Molecular Weight:282.34 g/mol
  • Melting Pt:164…165 °C
  • Storage Temperature:Ambient
  • EINECS:264-899-3
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications