Order Entry
Export
Orders LinkContactUsLinkComponent
17-AAG

17-AAG

Bottle, 10 mg
Catalog # APOSBIA4307-10MG
Irritant
Supplier:   Apollo Scientific
17-AAG
Catalog # APOSBIA4307-10MG
Supplier:  Apollo Scientific
CAS Number:  75747-14-7
17-AAG

Bottle, 10 mg

CAS Number:  75747-14-7
Price
102,00 €
Log in for availability
1 * 10 mg
Product Details & Documents

Hsp90 inhibitor.

Synonyms:  17-(Allylamino)-17-demethoxygeldanamycin, Tanespimycin, (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
  • CAS Number:75747-14-7
  • Formula:C₃₁H₄₃N₃O₈
  • MDL Number:MFCD04973892
  • Molecular Weight:585.7 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 mg

Specifications