Order Entry
Export
Orders LinkContactUsLinkComponent
Adenosine 99%

Adenosine 99%

Bottle, 100 g
Catalog # APOSBIA2545-100G
Irritant
Supplier:   Apollo Scientific
Adenosine 99%
Catalog # APOSBIA2545-100G
Supplier:  Apollo Scientific
CAS Number:  58-61-7
Adenosine 99%

Bottle, 100 g

CAS Number:  58-61-7
Price
86,90 €
Log in for availability
1 * 100 g
Product Details & Documents

Endogenous neurotransmitter.

Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Melting Pt:234…236 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-389-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications