Order Entry
Export
Orders LinkContactUsLinkComponent
Pipe Markers, Water

Pipe Markers, Water

Chilled water, Polyester, 250×26 mm
Catalog # BRDYN003412
Pipe Markers, Water
Catalog # BRDYN003412
Supplier:  
Pipe Markers, Water
Pipe Markers, Water

Chilled water, Polyester, 250×26 mm

Discontinued
Product Details & Documents

Individual markers on card with precut arrows, without pictogram.

  • Colour: White on green
  • Permanent acrylic
  • Pipe diameter: 20 to 49 mm
Specifications
  • Description:
    Chilled water
  • Dimensions:
    250×26 mm
  • Material:
    Polyester
  • Seq:
    9