Order Entry
Export
Orders LinkContactUsLinkComponent
 

Pipe Markers, Water

Condensed water, Polyester, 250×26 mm
  BRDYN003371
Pipe Markers, Water
  BRDYN003371
 :  
Pipe Markers, Water
Pipe Markers, Water

Condensed water, Polyester, 250×26 mm

 
 

Individual markers on card with precut arrows, without pictogram.

  • Colour: White on green
  • Permanent acrylic
  • Pipe diameter: 20 to 49 mm
 
  • Description:
    Condensed water
  • Dimensions:
    250×26 mm
  • Material:
    Polyester
  • Seq:
    8