Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Phusion® HF Buffer Pack - 6.0 ml 1 * 6 mL
Phusion® HF Buffer Pack - 6.0 ml 1 * 6 mL
Print
Phusion® HF Buffer Pack - 6.0 ml 1 * 6 mL
Catalog # NEWEB0518S
Supplier:
NEW ENGLAND BIOLABS
Phusion® HF Buffer Pack - 6.0 ml 1 * 6 mL
Catalog # NEWEB0518S
Supplier:
NEW ENGLAND BIOLABS
Expand image
Return to Top
Phusion® HF Buffer Pack - 6.0 ml 1 * 6 mL
Add to cart
Price
38,00 €
Log in
for availability
Unit of Measure
1 * 6 mL
-
+
Add to cart
Return Policy
Product Details & Documents