Order Entry
Export
Orders LinkContactUsLinkComponent
Chromatography columns with filter plates

Chromatography columns with filter plates

Chromatography column with PTFE tap, 80,00 mm, 400 mm, 29/32 NS
Catalog # NEUB1202-290800400
Chromatography columns with filter plates
Catalog # NEUB1202-290800400
Chromatography columns with filter plates

Chromatography column with PTFE tap, 80,00 mm, 400 mm, 29/32 NS

Price
538,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These chromatography columns feature a top sleeve with PTFE tap and are made from borosilicate glass 3.3.

Specifications
  • Ø int.:
    80,00 mm
  • Socket size:
    29/32 NS
  • Description:
    Chromatography column with PTFE tap
  • Length:
    400 mm
  • Seq:
    06