Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse IgM purification kit

Mouse IgM purification kit

LigaTrap® Mouse IgM purification resin, 1 KIT
Catalog # LIGALT-145
Mouse IgM purification kit
Catalog # LIGALT-145
CAS Number:  
Mouse IgM purification kit

LigaTrap® Mouse IgM purification resin, 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents

LigaTrap® Mouse IgM Purification Kit with LT technology utilises novel, patented ligands, designed for IgM antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of mouse IgM in just ten easy steps.

  • Designed to purify mouse IgM
  • For in vitro laboratory use
  • Binding capacity for mouse IgM is >10 mg/ml resin
  • Storage: 2 to 8 °C

LigaTrap® mouse IgM purification resin is engineered to purify high quality mouse IgM antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.

Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.

Delivery information: Each kit contains ten microspin columns prefilled with 0,1 ml of LigaTrap® mouse IgM purification resin, buffers and collection tubes.

Caution: Not for diagnostic or therapeutic use. Not for human or animal consumption.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    LigaTrap® Mouse IgM purification resin

Specifications