Various replacement parts
Line filter, stainless steel, LC-30AD, 228-35871-99, SHIM
Catalog # CTSA11-4129
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Line filter, stainless steel
- Seq:32
- OEM manufacturer:SHIM
- OEM equivalent:228-35871-99
- Model:LC-30AD