Orders LinkContactUsLinkComponent
Various replacement parts
Compression screw, 1,58 mm, stainless steel, 710, 712, 715, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, Alliance, WAT005070, WATE
Catalog # CTSACTS-12016
Supplier:   SCIENCIX
Various replacement parts
Catalog # CTSACTS-12016
Supplier:  SCIENCIX
Various replacement parts
Various replacement parts
Compression screw, 1,58 mm, stainless steel, 710, 712, 715, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, Alliance, WAT005070, WATE
Price
53,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
  • Description:
    Compression screw, 1,58 mm, stainless steel
  • Seq:
    11
  • OEM manufacturer:
    WATE
  • OEM equivalent:
    WAT005070
  • Model:
    710, 712, 715, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, Alliance