High Performance Lab Refrigerators, TSX
Product Details & Documents
Available in glass or solid door, the TSX series high-performance refrigerators combine advanced microprocessor temperature control with high-quality cabinet construction and alarm systems. Positive, forced-air circulation is designed to maintain temperature uniformity to protect important medical and pharmaceuticalgrade storage as well as laboratory media, reagents and more. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
- Forced-air circulation for temperature uniformity at all shelf levels
- Heat-free defrost for maximum temperature uniformity
- Quiet operation at just 52 dBA
- Four 2" casters for easy mobility; the front two are lockable
- Bright, LED interior lighting
- Keyed on/off switch with easy to use, setpoint security
- Self-closing door with 90° stop to assist with inventory loads
- Built-in remote alarm contacts
- Standard self powered 4 to 20 mA output
- Glass door versions are standard with triple pane glass door
- Designed to safely store media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade products requiring a temperature range of 2 to 8 °C
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor Speed is needed and periods of stability where the compressor runs at a lower speed
- Saving energy without compromising protection
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Standard door locks provide comfort and assurance
- Tested to GMP Clean Room Class A/ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP and hazardous drug compounding and other clean room environments
- Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863
V-drive technology: While conventional refrigerant, high-performance refrigerators and freezers use single speed compressors that continually cycle on and off, the TSX Series V-drive runs at variable speeds to adjust cooling performance to the conditions inside and outside of the refrigerator or freezer. When combined with the automated tuning control, this variable speed drive optimises the compressors running speed to the current conditions. When there are frequent door openings, or samples are added to the refrigerator or freezer, the control system detects the activity and increases the drive speed to bring temperatures back to the set point quickly. This innovative technology is one of the reasons the TSX Series delivers outstanding door opening recovery (DOR) speed, and more peace of mind for busy laboratories. When conditions are stable, such as when the unit is running overnight or on weekends, the drive runs at a low speed, reducing energy consumption while maintaining a stable temperature for the samples.
Ergonomically designed door handle with a soft-touch grip.The controller is angled to help prevent neck strain. Convenient, easy to clean and adjustable epoxy coated full and half shelves. Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Bright, LED interior lighting, self-closing multipaned glass or solid doors with 90° stop to assist with inventory loads, US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models*.
ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations. Manufactured in an award-winning, zero-waste to landfill facility, Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always - and anywhere - know the safety of your samples and the performance of your refrigerator or freezer. Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems. Alternate Door swing configuration and door types, Extra and Alternate wire baskets, shelves, and drawers, Free-standing or factory-installed chart recorders, Stainless Steel interior finish, E-Lock Adapter kits.
Certifications: UL, cUL, ENERGYSTAR, CE.
Specifications
- W×D×H ext.:711×962×1996 mm
- Capacity:650 L
- Description:Lab refrigerator, TSX2305SV
- Seq:02
- W×D×H int.:610×723×1473 mm
- Temperature:2...8 °C
- Shelves/drawers:4 shelves
- Refrigerant:R290
- Plug:CEE 7/7
- Door type:1 solid swing door
- Environmentally Preferable:Energy Efficient|Safer Human and Environmental Health
- Weight:165 kg
- Shipping weight:184 kg
- Cat. No.:HERATSX2305SV
- Specifications:4 casters; the front 2 are lockable
Bright, LED interior lighting
230 V, 16 A, 50 Hz - Alarm:Audible and visual
- Model:TSX2305SV
- Electrical:208-230 V, 50/60 Hz
- Display:Microprocessor
Services
Product Information
- Catalog #DescriptionAvailabilityPrice
- SERW1100.0100.01Installation of fridge at customer site. The device must be at the place of use. Functionality test and brief user introduction to functions. Travel fees included (cost of air travel excluded).Log in for availability1 * 1 Each - 610,00 €
- SERW1100.7000.01Installation Qualification and Operation Qualification of fridge at customer site. Qualification Protocols not included. Travel fees included (cost of air travel excluded).Log in for availability1 * 1 Each - 4 520,00 €
- SERW1100.1101.01Preventive maintenance, operational testing and calibration of fridge at customer site. 1× Temperature Calibration (1 probe). Calibration Certificate included. Spare parts and travel fees not includedLog in for availability1 * 1 Each - 776,00 €
- SERW1100.0900.01Preventive maintenance and operational testing of fridge at customer site. Spare parts and travel fees not includedLog in for availability1 * 1 Each - 300,00 €



