Pipette bulbs, Howorka-Ball®
50 ml, Black/red
Catalog # BEHRQL9273827
Supplier: BEHR
Jump to:
Product Details & Documents
These safety pipette bulbs are suitable for all standard tube diameters, including pipettes, burettes and filter tubes.
- No rigid connection with tube
- No valves or side vent on tubes
- No unstable air pockets
- Long operational life due to durable components
- Easily cleaned and steam sterilised if contaminated with liquids
Specifications
- Capacity:50 ml
- Colour:Black/red
- Seq:02