Order Entry
Export
Orders LinkContactUsLinkComponent
Magnetic stirring bars

Magnetic stirring bars

Magnetic stirring bar Ø 10×60 mm, 60 mm, 10 mm, AREC series
Catalog # VELPA00001061
Supplier:   VELP SCIENTIFIC
Magnetic stirring bars
Catalog # VELPA00001061
Supplier:  VELP SCIENTIFIC
Magnetic stirring bars
Magnetic stirring bars

Magnetic stirring bar Ø 10×60 mm, 60 mm, 10 mm, AREC series

Price
8,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Ø:
    10 mm
  • For:
    AREC series
  • Description:
    Magnetic stirring bar Ø 10×60 mm
  • Length:
    60 mm