Order Entry
Export
Orders LinkContactUsLinkComponent
 

Magnetic stirring bars

Magnetic stirring bar Ø 6×20 mm, 20 mm, 6 mm, AREC series
  VELPA00001057
Magnetic stirring bars
  VELPA00001057
Magnetic stirring bars
Magnetic stirring bars

Magnetic stirring bar Ø 6×20 mm, 20 mm, 6 mm, AREC series

 
3,50 €
   
1 * 1 items
 
 
  • Ø:
    6 mm
  • For:
    AREC series
  • Description:
    Magnetic stirring bar Ø 6×20 mm
  • Length:
    20 mm
  • Seq:
    06