WHEATON® Diagnostic Vials
Tubular vials, 10,00 ml, 22×55 mm, Borosilicate glass, Amber, No, 20-400
Catalog # WHEAW216376
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These vials are manufactured from borosilicate glass that conforms to USP Type I requirements. The screw thread design eliminates the need for crimping tools.
- Alternative to serum bottles and vials
- Ideal for lyophilisation
- Fits with I-Loc™ closure or screw cap with thin flange stopper
- Heavy walled glass provides impact resistance
Specifications
- Cap included:No
- Cap Size:20-400
- Ø×H:22×55 mm
- Capacity:10,00 ml
- Colour:Amber
- Description:Tubular vials
- Material:Borosilicate glass
- Seq:04



