Orders LinkContactUsLinkComponent
Robotic tips, Axygen®, for Beckman®
Robotic tips, Axygen®, for Beckman®
FX-20-R-S, 20 µl, +, -, Transparent, AP-96 / SPAN-8 / MP20, 50 racks, 96 each
Catalog # AXYGFX-20-R-S
   Low Manufacturing Impact
Supplier:   Corning
Robotic tips, Axygen®, for Beckman®
Catalog # AXYGFX-20-R-S
Supplier:  Corning
   Low Manufacturing Impact
Robotic tips, Axygen®, for Beckman®
Robotic tips, Axygen®, for Beckman®
FX-20-R-S, 20 µl, +, -, Transparent, AP-96 / SPAN-8 / MP20, 50 racks, 96 each
Price
444,00 €
Log in for availability
1 * 4.800 items
Product Details & Documents
Free of RNase, DNase, DNA and pyrogens.
  • Transparent PP
  • Sterile tips: Sterilised by gamma radiation
  • Filter tips: PE filter prevents any contamination of the pipette
  • Maxymum Recovery™ tips: Ultra-smooth surface prevents retention, loss of components, and denaturation of the sample
Specifications
  • Packed:
    50 racks, 96 each
  • Sterile:
    +
  • Volume:
    20 µl
  • Seq:
    25
  • Tip style:
    FX-20-R-S
  • Tip colour:
    Transparent
  • For use with:
    AP-96 / SPAN-8 / MP20
  • Filter:
    -
  • Cat. No.:
    AXYGFX-20-R-S
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact