Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Bar, length 1 m, Ø 22 mm, 2 thread connections M 12, steel powder coated, black 1 * 1 items
Catalog # LENZ06247613
Supplier:
Lenz Laborglas GmbH & CO.KG
Expand image
Bar, length 1 m, Ø 22 mm, 2 thread connections M 12, steel powder coated, black 1 * 1 items
Catalog # LENZ06247613
Supplier:
Lenz Laborglas GmbH & CO.KG