AOCS-Tintometer colour reference standards, Supelco®
AOCS-Tintometer Colour Reference Standard, 1.7R 12Y (5¼"), 500 mL
Catalog # SIAL134260-500ML
Supplier: Merck
AOCS-Tintometer colour reference standards, Supelco®
Catalog # SIAL134260-500ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
AOCS-Tintometer Colour Standard for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
- Certified under ISO 9001 Quality system.
- Full Traceability to International Standards.
- 500mL Pack Size supplied with MSDS
- 12 months shelf life.
- Tamper evident security packaging
Specifications
Specifications
- Pk:500 mL
- Description:AOCS-Tintometer Colour Reference Standard, 1.7R 12Y (5¼")



