Orders LinkContactUsLinkComponent
Fluid samplers, Liquid Master
Fluid samplers, Liquid Master
Fluid sampler, PTFE, 1500 ml, 2000 mm, 35 mm
Catalog # SAMP5035J-2000
Supplier:   Sampling Systems
Fluid samplers, Liquid Master
Catalog # SAMP5035J-2000
Supplier:  Sampling Systems
Fluid samplers, Liquid Master
Fluid samplers, Liquid Master
Fluid sampler, PTFE, 1500 ml, 2000 mm, 35 mm
Price
1 290,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The Liquid Master has been designed to be quickly stripped down for cleaning.
  • Quick and easy to use
  • Push button to press to take samples
  • For a larger volume, the sampler can be inserted deep into the product
Suitable for taking samples from low and medium viscosity liquids.
Specifications
  • Ø:
    35 mm
  • Capacity:
    1500 ml
  • Description:
    Fluid sampler
  • Length:
    2000 mm
  • Material:
    PTFE
  • Seq:
    07
Accessories
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0492
    Nylon
    15 - 30 mm
    Wire
    Log in for availability
    1 * 1 items - 55,50 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0514
    Nylon
    7 - 14 mm
    Wire
    Log in for availability
    1 * 1 items - 66,60 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0493
    Nylon
    15 - 30 mm
    Wire
    Log in for availability
    1 * 1 items - 66,60 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0494
    Nylon
    15 - 30 mm
    Wire
    Log in for availability
    1 * 1 items - 77,70 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0513
    Nylon
    7 - 14 mm
    Wire
    Log in for availability
    1 * 1 items - 55,50 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0495
    Nylon
    15 - 30 mm
    Wire
    Log in for availability
    1 * 1 items - 44,40 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0496
    Nylon
    30 - 45 mm
    Wire
    Log in for availability
    1 * 1 items - 55,50 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0497
    Nylon
    30 - 45 mm
    Wire
    Log in for availability
    1 * 1 items - 66,60 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0498
    Nylon
    30 - 45 mm
    Wire
    Log in for availability
    1 * 1 items - 77,70 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0499
    Nylon
    30 - 45 mm
    Wire
    Log in for availability
    1 * 1 items - 44,40 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0516
    Nylon
    7 - 14 mm
    Wire
    Log in for availability
    1 * 1 items - 59,30 €
Product Information
  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0515
    Nylon
    7 - 14 mm
    Wire
    Log in for availability
    1 * 1 items - 77,70 €