Microplates, 384-well, non binding
384-well microplate, non binding, flat bottom, Clear
Catalog # GREI781901_40
Supplier: Greiner Bio-One
Microplates, 384-well, non binding
Catalog # GREI781901_40
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
All plates are free from RNase, DNase, and pyrogens, and are also free of DNA polymerase inhibitors.
Specifications
- Colour:Clear
- Description:384-well microplate, non binding, flat bottom
- Seq:02
- Cat. No.:GREI781901_40
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact