Cryo.s Cryogenic Storage Tubes with Datamatrix
Product Details & Documents
Manufactured from high quality polypropylene, Cryo.s freezing tubes are suitable for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin such as blood, serum, and sperm, as well as antibodies, RNA, DNA, and protein samples.
- Sterile
- Endotoxin-free
- Free of RNase, DNase, and human DNA
- Non-pyrogenic and non-cytotoxic
These cryogenic sample tubes are manufactured from high quality polypropylene, which means they are ideally suited for use over a broad range of temperatures (–196 to 121 °C).
Laser etched, ECC200 code is the newest version of Datamatrix coding that is available on Greiner Bio One tubes. Datamatrix coding supports advanced error correction algorithms with capability to decipher partially damaged codes.
Compatible with automated liquid handling and de-/re-capping systems.
Caution: Cryo.s tubes are not intended for sample storage within reproductive medicine.
Documents
Certificates
Search by Lot NumberSpecifications
- Cap included:Yes
- Ø×H:12,4×83 mm
- Capacity:4,00 ml
- Description:Cryo.s Tube with external thread
- Cap colour:Natural
- Base:Round bottom, starfoot base
- Cat. No.:479-2445
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact



