ContactUsLinkComponent
The vacuette visio plus needle with optical venipuncture control can make an important contribution to simplifying blood collection routine for phlebotomists. the needle has a transparent plastic hub, which functions as a flash chamber. successful n 1 * 100 items
Catalog # GREI450040_100
Supplier:  Greiner Bio-One
The vacuette visio plus needle with optical venipuncture control can make an important contribution to simplifying blood collection routine for phlebotomists. the needle has a transparent plastic hub, which functions as a flash chamber. successful n 1 * 100 items
Catalog # GREI450040_100
Supplier:  Greiner Bio-One