Glass inserts, AQ™
Glass inserts, for use with (2 ml) 8 mm, (12×32 mm) opening screw top or 11 mm serum finish vials, 0,10 ml, Conical bottom, 5×31 mm, Glass, No
MICS9503S-05N
These vial inserts are made from borosilicate glass.
- Cap included:No
- Ø×H:5×31 mm
- Capacity:0,10 ml
- Description:Glass inserts, for use with (2 ml) 8 mm, (12×32 mm) opening screw top or 11 mm serum finish vials
- Material:Glass
- Seq:03
- Base:Conical bottom



