Glass inserts, AQ™
Glass inserts, self aligning, for 8 mm (2 ml) narrow mouth screw top or serum finish top vials, 0,20 ml, Conical bottom, 5×32 mm, Borosilicate glass, No
Catalog # MICS9502S-03E
Supplier: MICROSOLV
Jump to:
Product Details & Documents
These vial inserts are made from borosilicate glass.
Specifications
- Cap included:No
- Ø×H:5×32 mm
- Capacity:0,20 ml
- Description:Glass inserts, self aligning, for 8 mm (2 ml) narrow mouth screw top or serum finish top vials
- Material:Borosilicate glass
- Seq:07
- Base:Conical bottom



