Order Entry
Export
Orders LinkContactUsLinkComponent
4-(1,1-Difluoroethyl)acetophenone

4-(1,1-Difluoroethyl)acetophenone

Bottle, 1 g
Catalog # APOSPC53354-1G
Irritant
Supplier:   Apollo Scientific
4-(1,1-Difluoroethyl)acetophenone
Catalog # APOSPC53354-1G
Supplier:  Apollo Scientific
CAS Number:  1188932-40-2
4-(1,1-Difluoroethyl)acetophenone

Bottle, 1 g

CAS Number:  1188932-40-2
Price
869,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-[4-(1,1-Difluoroethyl)phenyl]ethanone
  • CAS Number:1188932-40-2
  • Formula:C₁₀H₁₀F₂O
  • MDL Number:MFCD26728226
  • Molecular Weight:184.183 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications