Order Entry
Export
Orders LinkContactUsLinkComponent
 

7-Hydroxyindole 95%

Glass bottle, 250 mg
Catalog # ACRO462292500
Corrosive
Irritant
7-Hydroxyindole 95%
Catalog # ACRO462292500
CAS Number:  2380-84-9
7-Hydroxyindole 95%
7-Hydroxyindole 95%

Glass bottle, 250 mg

CAS Number:  2380-84-9
Price
124,00 €
Log in for availability
1 * 250 mg
Product Details & Documents
  • CAS Number:2380-84-9
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152102
  • Molecular Weight:133.15 g/mol
  • Melting Pt:85…87 °C
  • Storage Temperature:Freezer
Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Solid or crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    83 to 88 °C
  • GC:
    ≥94.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg
  • Cat. No.:
    ACRO462292500

Specifications