7-Hydroxyindole 95%
Glass bottle, 1 g
Catalog # ACRO462290010
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:2380-84-9
- Formula:C₈H₇NO
- MDL Number:MFCD00152102
- Molecular Weight:133.15 g/mol
- Melting Pt:85…87 °C
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Appearance:Yellow to brown Solid or crystalline solid
- Infrared spectrum:Conforms
- Melting point:83 to 88 °C
- GC:≥94.0 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



