2-Methylbutane 99+%, extra pure
Glass bottle, 25 mL
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Catalog # ACRO126470250
Supplier: Thermo Fisher Scientific
CAS Number: 78-78-4
Jump to:
Product Details & Documents
Synonyms: iso-Pentane
- CAS Number:78-78-4
- Formula:CH₃CH₂CH(CH₃)₂
- MDL Number:MFCD00009338
- Molecular Weight:72.15 g/mol
- Boiling Pt:30 °C (760 mmHg)
- Melting Pt:–160 °C
- Density:0.62 g/cm³
- Flash Pt:–51 °C
- Storage Temperature:Refrigerator
- EINECS:201-142-8
- UN:1265
- ADR:3,I
Documents
ACRO12647
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3530 to 1.3550 (20°C, 589 nm)
- GC:≥99.0 %
- Residue after evaporation:≤10 ppm
Specifications
- Pack type:Glass bottle
- Pk:25 mL



